Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fabp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152961.3 | NP_694493.1 | 3 - 4 | 467 | 99 | |
GenBank mRNA | AF448057.1 | - | 3 - 4 | 364 | 99 | |
BC049060.1 | - | 3 - 4 | 467 | 99 | ||
CT725749.2 | - | 1 - 2 | 304 | 99 | ||
EH456910.1 | - | 3 - 4 | 496 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |