Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nitr3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198355.2 | NP_938169.2 | 3 - 4 | 474 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Danio rerio Inventoried Probe spans exons |