Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tcerg1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198357.3 | NP_938171.2 | 17 - 18 | 2623 | 93 | |
GenBank mRNA | BC056813.1 | - | 17 - 18 | 2639 | 93 | |
CN506442.1 | - | 3 - 4 | 466 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |