Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gria3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198360.2 | NP_938174.1 | 7 - 8 | 1106 | 87 | |
XM_005157132.3 | XP_005157189.1 | 7 - 8 | 1241 | 87 | ||
XM_005157133.3 | XP_005157190.1 | 7 - 8 | 1240 | 87 | ||
XM_009307719.3 | XP_009305994.1 | 7 - 8 | 1252 | 87 | ||
XM_021480987.1 | XP_021336662.1 | 7 - 8 | 1254 | 87 | ||
GenBank mRNA | AF525746.1 | - | 7 - 8 | 1106 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |