Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198817.2 | NP_942112.2 | 13 - 14 | 547 | 120 | |
XM_005159326.1 | XP_005159383.1 | 13 - 14 | 682 | 120 | ||
XM_005159327.4 | XP_005159384.1 | 12 - 13 | 614 | 120 | ||
GenBank mRNA | AY372242.1 | - | 13 - 14 | 562 | 120 | |
BC099983.1 | - | 13 - 14 | 562 | 120 | ||
BC122091.1 | - | 13 - 14 | 562 | 120 | ||
BC129155.1 | - | 13 - 14 | 547 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |