Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col1a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199214.1 | NP_954684.1 | 20 - 21 | 1306 | 135 | |
GenBank mRNA | AY380817.1 | - | 20 - 21 | 1306 | 135 | |
BC063249.1 | - | 20 - 21 | 1401 | 135 | ||
BC161663.1 | - | 20 - 21 | 1418 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |