Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199428.2 | NP_955460.2 | 4 - 5 | 466 | 116 | |
GenBank mRNA | BC053240.1 | - | 4 - 5 | 466 | 113 | |
BC164639.1 | - | 4 - 5 | 367 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |