Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199638.2 | NP_955932.2 | 4 - 5 | 875 | 79 | |
GenBank mRNA | BC049337.1 | - | 4 - 5 | 592 | 79 | |
EH519797.1 | - | 2 - 3 | 295 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |