Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200059.2 | NP_956353.1 | 5 - 6 | 694 | 103 | |
GenBank mRNA | BC059543.1 | - | 5 - 6 | 673 | 103 | |
BC078649.1 | - | 5 - 6 | 694 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |