Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200416.1 | NP_956710.1 | 7 - 8 | 884 | 85 | |
XM_005170010.4 | XP_005170067.1 | 8 - 9 | 1241 | 85 | ||
XM_009294204.3 | XP_009292479.1 | 8 - 9 | 1228 | 85 | ||
GenBank mRNA | BC054632.1 | - | 7 - 8 | 884 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |