Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ammecr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200581.1 | NP_956875.1 | 4 - 5 | 728 | 94 | |
GenBank mRNA | BC056704.1 | - | 4 - 5 | 728 | 94 | |
BC067667.1 | - | 4 - 5 | 764 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |