Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4e2rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200759.2 | NP_957053.1 | 4 - 5 | 570 | 66 | |
XM_017353532.2 | XP_017209021.1 | 4 - 5 | 573 | 66 | ||
GenBank mRNA | BC059582.1 | - | 4 - 5 | 519 | 66 | |
EH485715.1 | - | 4 - 5 | 563 | 66 | ||
KF294084.1 | - | 4 - 5 | 362 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |