Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200973.3 | NP_957267.1 | 5 - 6 | 956 | 56 | |
XM_005165131.3 | XP_005165188.1 | 5 - 6 | 577 | 56 | ||
XM_017356047.2 | XP_017211536.1 | 5 - 6 | 818 | 56 | ||
XM_021476040.1 | XP_021331715.1 | 6 - 7 | 590 | 56 | ||
GenBank mRNA | BC054126.1 | - | 5 - 6 | 541 | 56 | |
BC096776.1 | - | 5 - 6 | 957 | 56 | ||
BC164184.1 | - | 4 - 5 | 433 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |