Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205556.1 | NP_991119.1 | 3 - 4 | 375 | 67 | |
XM_005155593.4 | XP_005155650.1 | 3 - 4 | 353 | 67 | ||
GenBank mRNA | BC056527.1 | - | 3 - 4 | 305 | 67 | |
BC065617.1 | - | 3 - 4 | 375 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |