Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr2f6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205557.1 | NP_991120.1 | 3 - 4 | 756 | 109 | |
XM_005171224.4 | XP_005171281.1 | 3 - 4 | 1000 | 109 | ||
XM_021478933.1 | XP_021334608.1 | 3 - 4 | 894 | 109 | ||
GenBank mRNA | BC065330.1 | - | 3 - 4 | 756 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |