Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mknk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212723.1 | NP_997888.1 | 9 - 10 | 872 | 73 | |
GenBank mRNA | BC045391.1 | - | 9 - 10 | 872 | 73 | |
BC165349.1 | - | 8 - 9 | 690 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |