Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316890.1 | NP_001303819.1 | 4 - 5 | 781 | 91 | |
NM_212729.2 | NP_997894.2 | 4 - 5 | 469 | 91 | ||
XM_005158496.4 | XP_005158553.1 | 4 - 5 | 829 | 91 | ||
GenBank mRNA | BC054939.1 | - | 4 - 5 | 487 | 91 | |
BC067564.1 | - | 4 - 5 | 469 | 91 | ||
DT081919.1 | - | 4 - 5 | 618 | 91 | ||
EH433688.1 | - | 4 - 5 | 508 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |