Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: heg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212968.2 | NP_998133.2 | 7 - 8 | 2474 | 71 | |
GenBank mRNA | AY507658.1 | - | 7 - 8 | 2475 | 71 | |
AY507659.1 | - | 7 - 8 | 2475 | 71 | ||
AY507660.1 | - | 7 - 8 | 2475 | 71 | ||
BC162109.1 | - | 7 - 8 | 2473 | 71 | ||
BC162480.1 | - | 7 - 8 | 2473 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |