Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212988.1 | NP_998153.1 | 3 - 4 | 448 | 71 | |
XM_005159885.4 | XP_005159942.1 | 4 - 5 | 689 | 71 | ||
GenBank mRNA | BC054680.1 | - | 3 - 4 | 448 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |