Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212988.1 | NP_998153.1 | 9 - 10 | 1223 | 85 | |
XM_005159885.4 | XP_005159942.1 | 10 - 11 | 1464 | 85 | ||
GenBank mRNA | BC054680.1 | - | 9 - 10 | 1223 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |