Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kpna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213070.1 | NP_998235.1 | 7 - 8 | 1276 | 147 | |
GenBank mRNA | BC044523.1 | - | 7 - 8 | 1276 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |