Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc35e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213074.1 | NP_998239.1 | 5 - 6 | 959 | 93 | |
GenBank mRNA | BC046896.1 | - | 5 - 6 | 959 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |