Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbwd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213253.1 | NP_998418.1 | 14 - 15 | 1016 | 79 | |
GenBank mRNA | BC065429.1 | - | 14 - 15 | 1016 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |