Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmed10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001310040.1 | NP_001296969.1 | 4 - 5 | 584 | 75 | |
NM_214698.2 | NP_999863.1 | 4 - 5 | 590 | 75 | ||
GenBank mRNA | BC059496.1 | - | 4 - 5 | 561 | 75 | |
BC067575.1 | - | 4 - 5 | 556 | 75 | ||
EH541878.1 | - | 4 - 5 | 603 | 75 | ||
EH611403.1 | - | 1 - 2 | 453 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |