Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281986.1 | NP_001268915.1 | 9 - 10 | 1094 | 61 | |
NM_214700.2 | NP_999865.1 | 10 - 11 | 1238 | 61 | ||
XM_021476786.1 | XP_021332461.1 | 10 - 11 | 1204 | 61 | ||
GenBank mRNA | BC051155.1 | - | 10 - 11 | 1186 | 61 | |
BC067579.1 | - | 10 - 11 | 1171 | 61 | ||
EH611446.1 | - | 1 - 2 | 303 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |