Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281986.1 | NP_001268915.1 | 6 - 7 | 733 | 103 | |
NM_214700.2 | NP_999865.1 | 7 - 8 | 877 | 103 | ||
XM_021476786.1 | XP_021332461.1 | 7 - 8 | 843 | 103 | ||
GenBank mRNA | BC051155.1 | - | 7 - 8 | 825 | 103 | |
BC067579.1 | - | 7 - 8 | 810 | 103 | ||
CT720868.1 | - | 6 - 7 | 673 | 103 | ||
EH611446.1 | - | 4 - 5 | 664 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |