Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhej1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214749.1 | NP_999914.1 | 3 - 4 | 564 | 100 | |
GenBank mRNA | BC068348.1 | - | 3 - 4 | 564 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |