Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trmt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077281.1 | NP_001070749.1 | 10 | 1857 | 158 | |
XM_017356915.2 | XP_017212404.1 | 10 | 1920 | 158 | ||
GenBank mRNA | BC124659.1 | - | 10 | 1857 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |