Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult3st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079947.1 | NP_001073416.1 | 6 | 1042 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |