Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc43a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083000.1 | NP_001076469.1 | 15 | 3453 | 136 | |
GenBank mRNA | BC133091.1 | - | 15 | 3453 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Inventoried Both primers and probe map within a single exon |