Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008733.1 | NP_001008733.1 | 8 | 1047 | 129 | |
XM_021474253.1 | XP_021329928.1 | 9 | 1115 | 129 | ||
GenBank mRNA | BC086734.1 | - | 8 | 1047 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |