Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gxylt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110200.1 | NP_001103670.1 | 7 | 1651 | 111 | |
GenBank mRNA | BC134947.1 | - | 7 | 1651 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |