Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pex7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013532.2 | NP_001013550.1 | 10 | 1252 | 95 | |
GenBank mRNA | BC090898.1 | - | 10 | 1254 | 95 | |
BC154272.1 | - | 10 | 1223 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |