Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh2aa15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB160960.1 | - | 1 | 48 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |