Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh2aa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009587.2 | NP_001009587.2 | 1 | 1548 | 121 | |
GenBank mRNA | AB160959.1 | - | 1 | 1548 | 121 |
Gene Symbol: pcdh2aa15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB160958.1 | - | 1 | 1548 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |