Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fancf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045234.2 | NP_001038699.2 | 1 | 1356 | 91 | |
GenBank mRNA | BC139581.1 | - | 1 | 1356 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |