Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shisa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080662.2 | NP_001074131.2 | 1 | 461 | 69 | |
GenBank mRNA | BC129191.1 | - | 1 | 452 | 69 | |
EH594153.1 | - | 1 | 489 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |