Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vangl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153674.1 | NP_705960.1 | 4 | 951 | 113 | |
GenBank mRNA | AF428249.1 | - | 4 | 951 | 113 | |
BC065983.1 | - | 1042 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |