Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199620.1 | NP_955914.1 | 8 | 1247 | 93 | |
GenBank mRNA | AY648766.1 | - | 8 | 1246 | 93 | |
BC056822.1 | - | 8 | 1247 | 93 | ||
BC153472.1 | - | 3 | 606 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |