Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc43a2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200251.2 | NP_956545.1 | 5 | 3041 | 137 | |
GenBank mRNA | BC047854.1 | - | 5 | 3042 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |