Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: becn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200872.1 | NP_957166.1 | 5 | 349 | 78 | |
GenBank mRNA | AB266448.1 | - | 4 | 306 | 78 | |
BC063319.1 | - | 5 | 349 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |