Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213041.3 | NP_998206.1 | 2156 | 103 | ||
GenBank mRNA | BC064707.4 | - | 2156 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio |