Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ciao2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002449.1 | NP_001002449.1 | 1 - 2 | 198 | 96 | |
GenBank mRNA | BC076022.1 | - | 1 - 2 | 198 | 96 | |
BC162658.1 | - | 1 - 2 | 160 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |