Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ciao2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002449.1 | NP_001002449.1 | 4 - 5 | 450 | 66 | |
GenBank mRNA | BC076022.1 | - | 4 - 5 | 450 | 66 | |
BC155174.1 | - | 4 - 5 | 454 | 66 | ||
BC155260.1 | - | 4 - 5 | 474 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |