Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmtm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002492.1 | NP_001002492.1 | 2 - 3 | 698 | 73 | |
GenBank mRNA | BC076315.1 | - | 2 - 3 | 698 | 73 | |
BC152629.1 | - | 2 - 3 | 841 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |