Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab11bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002555.1 | NP_001002555.1 | 4 - 5 | 709 | 90 | |
GenBank mRNA | BC076247.1 | - | 4 - 5 | 709 | 90 | |
BC152630.1 | - | 4 - 5 | 673 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |