Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atxn7l2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002655.2 | NP_001002655.2 | 4 - 5 | 748 | 109 | |
GenBank mRNA | BC076493.1 | - | 4 - 5 | 660 | 109 | |
BC096979.1 | - | 4 - 5 | 701 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |