Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp1r14bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002692.1 | NP_001002692.1 | 1 - 2 | 694 | 115 | |
GenBank mRNA | BC076513.1 | - | 1 - 2 | 694 | 115 | |
BC165900.1 | - | 1 - 2 | 232 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |