Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002708.1 | NP_001002708.1 | 3 - 4 | 462 | 146 | |
GenBank mRNA | BC076096.1 | - | 3 - 4 | 462 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |