Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh3bgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003524.2 | NP_001003524.1 | 9 - 10 | 654 | 85 | |
XM_005169682.4 | XP_005169739.1 | 10 - 11 | 823 | 85 | ||
XM_005169686.4 | XP_005169743.1 | 9 - 10 | 708 | 85 | ||
XM_005169688.4 | XP_005169745.1 | 8 - 9 | 561 | 85 | ||
GenBank mRNA | BC077137.1 | - | 9 - 10 | 627 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |